Gene Transcripts

Gene Symbol
Tm6sf2
Entrez Gene ID
107770
Gene Name
transmembrane 6 superfamily member 2
Gene Aliases
Chromosome Location
Chr. 8: 70072924 - 70080066 on Build GRCm38
UniGene ID
Mm.37974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293795.1 NP_001280724.1 8 823
NM_181540.4 NP_853518.1 8 817
GenBank mRNA AK077126.1 - 8 810
BC024498.1 - 8 811 4982620

View Full Product Details