Gene Transcripts

Gene Symbol
Fbxw17
Entrez Gene ID
109082
Gene Name
F-box and WD-40 domain protein 17
Gene Aliases
1110064L07Rik, C85886
Chromosome Location
Chr. 13: 50417834 - 50433781 on Build GRCm38
UniGene ID
Mm.18638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175401.3 NP_780610.2 10 1375
GenBank mRNA AK075676.1 - 2 1500
AK135553.1 - 8 2227
AK155157.1 - 11 1889
BC040428.1 - 10 1375 3327345

View Full Product Details