Gene Transcripts

Gene Symbol
Evx2
Entrez Gene ID
14029
Gene Name
even skipped homeotic gene 2 homolog
Gene Aliases
Evx-2
Chromosome Location
Chr. 2: 74655524 - 74661518 on Build GRCm38
UniGene ID
Mm.25365
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007967.2 NP_031993.1 1, 2 419
GenBank mRNA BC061467.1 - 557
M93128.1 - 1, 2 419

View Full Product Details