Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022167.3 | NP_071450.2 | 11 | 2829 | |
NR_110010.1 | 10 | 2633 | |||
XM_005257572.4 | XP_005257629.1 | 11 | 3143 | ||
GenBank mRNA | AJ277442.1 | - | 11 | 2829 | |
AL833210.1 | - | 1 | 1448 | ||
AY358090.1 | - | 10 | 2529 | ||
BC052262.1 | - | 11 | 2749 | 6380013 |