Gene Transcripts

Gene Symbol
SGPP1
Entrez Gene ID
81537
Gene Name
sphingosine-1-phosphate phosphatase 1
Gene Aliases
SPPase1
Chromosome Location
Chr. 14: 63684216 - 63728095 on Build GRCh38
UniGene ID
Hs.24678
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030791.3 NP_110418.1 3 1117
XM_017021678.1 XP_016877167.1 4 1181
GenBank mRNA AJ293294.1 - 3 998
AK314188.1 - 3 1063
BC063839.1 - 3 1060 6184825
BC070060.1 - 3 1077 30347615

View Full Product Details