Gene Transcripts

Gene Symbol
MYB
Entrez Gene ID
4602
Gene Name
MYB proto-oncogene, transcription factor
Gene Aliases
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location
Chr. 6: 135180981 - 135219173 on Build GRCh38
UniGene ID
Hs.606320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130172.1 NP_001123644.1 11 1624
NM_001130173.1 NP_001123645.1 12 1996
NM_001161656.1 NP_001155128.1 12 1987
NM_001161657.1 NP_001155129.1 10 1378
NM_001161658.1 NP_001155130.1 12 1948
NM_001161659.1 NP_001155131.1 11 1633
NM_001161660.1 NP_001155132.1 10 1528
NM_005375.3 NP_005366.2 11 1633
NR_134958.1 12 1755
NR_134959.1 12 1746
NR_134960.1 12 1689
NR_134961.1 12 1816
NR_134962.1 11 1534
NR_134963.1 11 1633
NR_134964.1 12 1680
NR_134965.1 13 1811
GenBank mRNA AF104863.1 - 11 1511
AJ606317.1 - 12 1755
AJ606318.1 - 12 1816
AJ606319.1 - 12 1996
AJ606320.1 - 11 1624
AJ606321.1 - 12 1746
AJ606322.1 - 12 1689
AJ606323.1 - 11 1633
AJ606324.1 - 11 1633
AJ616235.1 - 11 1548
AJ616801.1 - 1 88
AY787443.1 - 11 1434
AY787444.1 - 9 1141
AY787445.1 - 11 1484
AY787446.1 - 11 1335
AY787447.1 - 10 1329
AY787448.1 - 10 1179
AY787449.1 - 11 1235
AY787450.1 - 12 1490
AY787451.1 - 11 1434
AY787452.1 - 11 1434
AY787453.1 - 11 1434
AY787454.1 - 12 1556
AY787455.1 - 13 1928
AY787456.1 - 13 1612
AY787457.1 - 13 1527
AY787458.1 - 12 1481
AY787459.1 - 12 1547
AY787460.1 - 13 1910
AY787461.1 - 12 1788
AY787462.1 - 12 1937
AY787463.1 - 12 1703
AY787464.1 - 12 1617
AY787465.1 - 12 2031
AY787466.1 - 13 1673
AY787467.1 - 12 1797
AY787468.1 - 12 1749
AY787469.1 - 13 1805
AY787470.1 - 11 1349
AY787471.1 - 12 1405
AY787472.1 - 12 1532
AY787473.1 - 12 1946
AY787474.1 - 12 1712
AY787475.1 - 11 1434
BC035996.1 - 5 664
BC064955.1 - 11 1585 6069320
M13665.1 - 9 1296
M15024.1 - 11 1547
X52125.1 - 11 1622
X52126.1 - 11 1550

View Full Product Details