Gene Transcripts

Gene Symbol
TM2D1
Entrez Gene ID
83941
Gene Name
TM2 domain containing 1
Gene Aliases
BBP
Chromosome Location
Chr. 1: 61681047 - 61725423 on Build GRCh38
UniGene ID
Hs.656790
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032027.2 NP_114416.1 5 793
NR_135160.1 5 793
NR_135162.1 5 816
GenBank mRNA AF353990.1 - 5 793
AK311444.1 - 5 439
AK311705.1 - 5 440
BC029486.1 - 5 511 5271098
BC048995.1 - 5 509
BG700625.1 - 5 507
BU664262.1 - 5 460

View Full Product Details