Gene Transcripts

Gene Symbol
HNRNPLL
Entrez Gene ID
92906
Gene Name
heterogeneous nuclear ribonucleoprotein L like
Gene Aliases
HNRPLL, SRRF
Chromosome Location
Chr. 2: 38563186 - 38603036 on Build GRCh38
UniGene ID
Hs.445497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142650.1 NP_001136122.1 6 823
NM_138394.3 NP_612403.2 5 1079
XM_005264639.3 XP_005264696.2 5 1076
XM_005264640.3 XP_005264697.2 6 822
XM_011533165.2 XP_011531467.1 5 1076
XM_011533166.2 XP_011531468.1 5 1076
GenBank mRNA AK000155.1 - 6 727
AK291462.1 - 5 944
AK292259.1 - 6 816
AL512692.1 - 4 473
AY236963.2 - 6 668
BC008217.1 - 6 716 3449607
BC017480.1 - 5 925 3502860
KU178839.1 - 5 669
KU178840.1 - 5 669
KU178841.1 - 5 669

View Full Product Details