Gene Transcripts

Gene Symbol
PHF21B
Entrez Gene ID
112885
Gene Name
PHD finger protein 21B
Gene Aliases
BHC80L, PHF4
Chromosome Location
Chr. 22: 44881162 - 45009928 on Build GRCh38
UniGene ID
Hs.254097
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135862.2 NP_001129334.1 14 2499
NM_001242450.1 NP_001229379.1 13 2539
NM_001284296.1 NP_001271225.1 14 2585
NM_138415.4 NP_612424.1 13 2670
NR_134535.1 12 2431
XM_006724122.1 XP_006724185.1 13 2679
GenBank mRNA AK091480.1 - 14 2585
AK092243.1 - 14 2499
AK095777.1 - 7 1651
BC012187.1 - 13 2670 4634631
CN311600.1 - 1 24

View Full Product Details