Gene Transcripts

Gene Symbol
YIPF6
Entrez Gene ID
286451
Gene Name
Yip1 domain family member 6
Gene Aliases
FinGER6
Chromosome Location
Chr. X: 68498374 - 68572413 on Build GRCh38
UniGene ID
Hs.82719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195214.1 NP_001182143.1 6 1192
NM_173834.3 NP_776195.2 7 1321
XM_011530924.1 XP_011529226.1 7 1103
GenBank mRNA AK024673.1 - 3 627
AK303992.1 - 6 922
BC012469.1 - 7 1059 4452699

View Full Product Details