Gene Transcripts

Gene Symbol
ZNF410
Entrez Gene ID
57862
Gene Name
zinc finger protein 410
Gene Aliases
APA-1, APA1
Chromosome Location
Chr. 14: 73886615 - 73932288 on Build GRCh38
UniGene ID
Hs.270869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242924.1 NP_001229853.1 5 851
NM_001242926.1 NP_001229855.1 4 800
NM_001242928.1 NP_001229857.1 4 800
NM_021188.2 NP_067011.1 4 800
NR_040251.1 4 768
GenBank mRNA AK223261.1 - 4 556
AK293350.1 - 5 607
AK298307.1 - 5 523
AK298520.1 - 4 576
AK301382.1 - 4 453
AK309957.1 - 5 596
AK310031.1 - 3 342
AK315206.1 - 4 558
BC004357.1 - 4 544
BC050683.1 - 4 610 5752452
U90919.1 - 4 532

View Full Product Details