Gene Transcripts
- Gene Symbol
- TARDBP
- Entrez Gene ID
- 23435
- Gene Name
- TAR DNA binding protein
- Gene Aliases
- ALS10, TDP-43
- Chromosome Location
- Chr. 1: 11012622 - 11030528 on Build GRCh38
- UniGene ID
- Hs.300624
- Species
- Homo sapiens
- Gene Symbol
- LOC643387
- Entrez Gene ID
- 643387
- Gene Name
- TAR DNA binding protein pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 2: 238231686 - 238234345 on Build GRCh38
- UniGene ID
- Hs.598359
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026923.1 |
|
1 |
1962 |
|
View Full Product Details