Gene Transcripts

Gene Symbol
TEK
Entrez Gene ID
7010
Gene Name
TEK receptor tyrosine kinase
Gene Aliases
CD202B, TIE-2, TIE2, VMCM, VMCM1
Chromosome Location
Chr. 9: 27109141 - 27230178 on Build GRCh38
UniGene ID
Hs.89640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000459.4 NP_000450.2 12 2200
NM_001290077.1 NP_001277006.1 11 2071
NM_001290078.1 NP_001277007.1 10 1759
XM_005251561.1 XP_005251618.1 12 1867
XM_005251563.1 XP_005251620.1 11 1738
GenBank mRNA AB086825.1 - 11 1727
AB208796.1 - 12 1878
AK291775.1 - 12 1888
AK294887.1 - 10 1760
AK295043.1 - 11 1797
BC035514.1 - 12 1875 5228999
L06139.1 - 12 1898

View Full Product Details