Gene Transcripts

Gene Symbol
CDH12
Entrez Gene ID
1010
Gene Name
cadherin 12
Gene Aliases
CDHB
Chromosome Location
Chr. 5: 21750191 - 22853622 on Build GRCh38
UniGene ID
Hs.113684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317227.1 NP_001304156.1 2 596
NM_001317228.1 NP_001304157.1 3 711
NM_004061.4 NP_004052.2 5 1225
XM_011513927.2 XP_011512229.1 3 502
XM_017008920.1 XP_016864409.1 4 852
XM_017008921.1 XP_016864410.1 5 945
GenBank mRNA AK295123.1 - 3 711
AK314800.1 - 2 596
BC047608.1 - 5 930 5751948
L33477.1 - 5 1226
L34057.1 - 1 231

View Full Product Details