Gene Transcripts

Gene Symbol
CRY1
Entrez Gene ID
1407
Gene Name
cryptochrome circadian clock 1
Gene Aliases
PHLL1
Chromosome Location
Chr. 12: 106991364 - 107093872 on Build GRCh38
UniGene ID
Hs.151573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004075.4 NP_004066.1 5 1553
XM_017018832.1 XP_016874321.1 6 1688
GenBank mRNA AK125915.1 - 7 1216
AK290552.1 - 5 853
BC017466.1 - 5 976
BC030519.1 - 5 1226 5169081
D83702.1 - 5 1243
D84657.1 - 5 657

View Full Product Details