Gene Transcripts

Gene Symbol
RASGEF1A
Entrez Gene ID
221002
Gene Name
RasGEF domain family member 1A
Gene Aliases
CG4853
Chromosome Location
Chr. 10: 43194533 - 43266919 on Build GRCh38
UniGene ID
Hs.125293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282862.1 NP_001269791.1 10 1323
NM_145313.3 NP_660356.2 10 1295
XM_005271809.3 XP_005271866.1 9 1173
XM_011539500.2 XP_011537802.1 9 1283
GenBank mRNA AK095136.1 - 10 1323
AK127432.1 - 10 3721
BC022548.1 - 10 1295 4814750

View Full Product Details