Gene Transcripts

Gene Symbol
Slc28a2
Entrez Gene ID
269346
Gene Name
solute carrier family 28 (sodium-coupled nucleoside transporter), member 2
Gene Aliases
2010208B10Rik, B430217P18, BB152493, Cnt2
Chromosome Location
Chr. 2: 122426355 - 122461152 on Build GRCm38
UniGene ID
Mm.29510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172980.2 NP_766568.1 17 3009
GenBank mRNA AF079853.2 - 16 1884
AK080957.1 - 17 3010
AK152776.1 - 16 1944
BC055376.1 - 16 1903 30077988

Gene Symbol
Gm14085
Entrez Gene ID
381417
Gene Name
predicted gene 14085
Gene Aliases
OTTMUSG00000015615
Chromosome Location
Chr. 2: 122484941 - 122528040 on Build GRCm38
UniGene ID
Mm.482295
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085518.1 NP_001078987.1 16 2847

View Full Product Details