Gene Transcripts

Gene Symbol
GDF10
Entrez Gene ID
2662
Gene Name
growth differentiation factor 10
Gene Aliases
BMP-3b, BMP3B
Chromosome Location
Chr. 10: 47300224 - 47313577 on Build GRCh38
UniGene ID
Hs.2171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004962.3 NP_004953.1 1 678
GenBank mRNA AK310360.1 - 1 535
AK315322.1 - 1 534
D49492.1 - 1 516

View Full Product Details