Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010174.1 | NP_034304.1 | 2 | 118 | |
GenBank mRNA | AK142156.1 | - | 2 | 156 | |
BC002082.1 | - | 2 | 148 | 3485639 | |
BC089542.1 | - | 2 | 156 | 30300963 | |
U02883.1 | - | 2 | 118 | ||
X14961.1 | - | 2 | 139 |