Gene Transcripts

Gene Symbol
E2f1
Entrez Gene ID
13555
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, mKIAA4009
Chromosome Location
Chr. 2: 154559400 - 154569892 on Build GRCm38
UniGene ID
Mm.18036
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291105.1 NP_001278034.1 7 1352
NM_007891.5 NP_031917.1 7 1396
GenBank mRNA AF483514.1 - 7 1238
AF483515.1 - 7 1238
AK017841.1 - 7 1344
AK153968.1 - 7 1361
AK220226.1 - 7 1093
BC052160.1 - 7 1338 6330425
L21973.1 - 7 1356

View Full Product Details