Gene Transcripts

Gene Symbol
Cyp2c38
Entrez Gene ID
13097
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 38
Gene Aliases
Chromosome Location
Chr. 19: 39389556 - 39463089 on Build GRCm38
UniGene ID
Mm.42100
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010002.3 NP_034132.2 9 1984
GenBank mRNA AK050051.1 - 9 1985

View Full Product Details