Gene Transcripts

Gene Symbol
Olfr215
Entrez Gene ID
258438
Gene Name
olfactory receptor 215
Gene Aliases
MOR119-2
Chromosome Location
Chr. 6: 116582012 - 116582944 on Build GRCm38
UniGene ID
Mm.357743
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146446.1 NP_666657.1 1 487
GenBank mRNA BC125561.1 - 1 574
BC125563.1 - 1 574 40130078

View Full Product Details