Gene Transcripts

Gene Symbol
Sucla2
Entrez Gene ID
20916
Gene Name
succinate-Coenzyme A ligase, ADP-forming, beta subunit
Gene Aliases
4930547K18Rik
Chromosome Location
Chr. 14: 73552786 - 73596144 on Build GRCm38
UniGene ID
Mm.38951
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011506.3 NP_035636.1 2 250
GenBank mRNA AF058955.1 - 1 117
AK005273.1 - 2 232
AK081965.1 - 2 271
AK088801.1 - 2 251
AK132762.1 - 2 240
AK150594.1 - 2 250
AK152783.1 - 2 250
AK153198.1 - 2 250
AK160130.1 - 2 251
AK160652.1 - 2 239
BC056353.1 - 2 241 6808360
BC057605.1 - 2 241 6808360

View Full Product Details