Gene Transcripts
- Gene Symbol
- Ap5z1
- Entrez Gene ID
- 231855
- Gene Name
- adaptor-related protein complex 5, zeta 1 subunit
- Gene Aliases
- C330006K01Rik
- Chromosome Location
- Chr. 5: 142463205 - 142478716 on Build GRCm38
- UniGene ID
- Mm.78250
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172725.2 |
NP_766313.2
|
15, 16 |
2094 |
|
GenBank mRNA |
AK040924.1 |
- |
13, 14 |
1646 |
|
AK082759.1 |
- |
7, 8 |
921 |
|
AK129135.1 |
- |
12 |
3292 |
|
AK146431.1 |
- |
15, 16 |
1972 |
|
AK152402.1 |
- |
15, 16 |
1972 |
|
AK153787.1 |
- |
14 |
3613 |
|
AK185767.1 |
- |
2, 3 |
156 |
|
AK187973.1 |
- |
2, 3 |
157 |
|
AK215647.1 |
- |
2, 3 |
156 |
|
BC057548.1 |
- |
15, 16 |
1901 |
|
BC144755.1 |
- |
13, 14 |
1577 |
|
View Full Product Details