Gene Transcripts

Gene Symbol
Nrbp1
Entrez Gene ID
192292
Gene Name
nuclear receptor binding protein 1
Gene Aliases
B230344L17Rik, Nrbp
Chromosome Location
Chr. 5: 31228089 - 31251566 on Build GRCm38
UniGene ID
Mm.292040
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147201.2 NP_671734.1 3 331
GenBank mRNA AF302138.1 - 3 330
AK046142.1 - 3 325
AK077440.1 - 3 367
AK170501.1 - 3 347
AK191367.1 - 4 211
AK217229.1 - 2 258
BC004756.1 - 3 301 3154089
BC018463.1 - 3 305 3598438

View Full Product Details