Gene Transcripts

Gene Symbol
Tmc7
Entrez Gene ID
209760
Gene Name
transmembrane channel-like gene family 7
Gene Aliases
1700030H01Rik, C230064B05, C630024K23Rik
Chromosome Location
Chr. 7: 118535841 - 118584790 on Build GRCm38
UniGene ID
Mm.69380
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172476.4 NP_766064.2 2 172
GenBank mRNA AK082562.1 - 2 201
AK083189.1 - 2 175
AK132820.1 - 2 201
AY236499.1 - 2 233
AY263161.1 - 2 201

View Full Product Details