Gene Transcripts

Gene Symbol
1700016D06Rik
Entrez Gene ID
76413
Gene Name
RIKEN cDNA 1700016D06 gene
Gene Aliases
Chromosome Location
Chr. 8: 11654921 - 11678750 on Build GRCm38
UniGene ID
Mm.440041
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024271.1 NP_077233.1 4 366
GenBank mRNA AK006019.1 - 4 339
AY026045.1 - 4 366
BC048664.1 - 4 358 6773850

View Full Product Details