Gene Transcripts

Gene Symbol
Arsg
Entrez Gene ID
74008
Gene Name
arylsulfatase G
Gene Aliases
6330406P08Rik, AI846872
Chromosome Location
Chr. 11: 109450845 - 109573330 on Build GRCm38
UniGene ID
Mm.482224
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028710.3 NP_082986.3 7 1385
GenBank mRNA AK018132.2 - 4 802
AK158726.1 - 7 1386
AK173082.1 - 4 484
AK192752.1 - 3 133
BC022158.1 - 3 215
BC084731.1 - 4 1075 6813261

View Full Product Details