Gene Transcripts

Gene Symbol
Ears2
Entrez Gene ID
67417
Gene Name
glutamyl-tRNA synthetase 2 (mitochondrial)(putative)
Gene Aliases
3230401I01Rik, mKIAA1970
Chromosome Location
Chr. 7: 122038654 - 122067185 on Build GRCm38
UniGene ID
Mm.45171
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK014324.2 - 862
AK129469.1 - 828
AK167506.1 - 4 874
AK169225.1 - 4 863
BC025907.1 - 4 880 5064949

View Full Product Details