Gene Transcripts

Gene Symbol
Fbxo8
Entrez Gene ID
50753
Gene Name
F-box protein 8
Gene Aliases
Fbx8
Chromosome Location
Chr. 8: 56551134 - 56593939 on Build GRCm38
UniGene ID
Mm.251174
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015791.3 NP_056606.2 2 433
GenBank mRNA AF176527.1 - 2 440
AK002538.1 - 2 444
AK012401.1 - 2 451
AK029989.1 - 2 434
AK077595.1 - 2 455
AK078111.1 - 2 453
AK147748.1 - 2 450
AK160145.1 - 2 414
AK162592.1 - 2 450
BC011100.1 - 2 454 4011855
BC034854.1 - 2 365 3468704

View Full Product Details