Gene Transcripts

Gene Symbol
Rabep1
Entrez Gene ID
54189
Gene Name
rabaptin, RAB GTPase binding effector protein 1
Gene Aliases
Rab5ep, rabaptin-5, rabaptin-5alpha
Chromosome Location
Chr. 11: 70844723 - 70943105 on Build GRCm38
UniGene ID
Mm.7087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291141.1 NP_001278070.1 3 546
NM_001291142.1 NP_001278071.1 4 675
NM_001291143.1 NP_001278072.1 5 746
NM_019400.3 NP_062273.2 4 675
GenBank mRNA AB001750.1 - 5 582
AF248489.1 - 4 461
AF248490.1 - 3 332
AK019413.1 - 4 649
AK134587.1 - 4 674
AK137236.1 - 4 640
AK151901.1 - 4 644
BC009814.1 - 4 671
BC060166.1 - 4 654
BC080274.1 - 4 648
BC129853.1 - 4 494 40109688
BC129854.1 - 4 494 40109689
D86066.1 - 4 511

View Full Product Details