Gene Transcripts

Gene Symbol
Tspan6
Entrez Gene ID
56496
Gene Name
tetraspanin 6
Gene Aliases
6720473L21Rik, AI316786, T245, TSPAN-6, Tm4sf, Tm4sf6
Chromosome Location
Chr. X: 133891070 - 133898429 on Build GRCm38
UniGene ID
Mm.46701
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019656.3 NP_062630.2 5 633
GenBank mRNA AF053454.1 - 5 580
AK146673.1 - 5 538
AK149204.1 - 5 543
AK160313.1 - 5 527
AK167384.1 - 5 529
BC003733.1 - 5 567 3590862
BC110314.1 - 5 542 5344878

View Full Product Details