Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026187.5 | NP_080463.3 | 8, 9 | 1564 | |
GenBank mRNA | AK004948.2 | - | 6, 7 | 1994 | |
AK012672.1 | - | 1, 2 | 137 | ||
AK156255.1 | - | 8, 9 | 1526 | ||
AK157405.1 | - | 8, 9 | 1526 | ||
BC039910.1 | - | 6, 7 | 2074 | ||
BC057896.1 | - | 3, 4 | 406 | ||
BC071240.1 | - | 8, 9 | 1494 | ||
BC150879.1 | - | 8, 9 | 1514 | 9087790 |