Gene Transcripts
- Gene Symbol
- Birc5
- Entrez Gene ID
- 64041
- Gene Name
- baculoviral IAP repeat-containing 5
- Gene Aliases
- AP14
- Chromosome Location
- Chr. 10: 106856097 - 106864419 on Build Rnor_6.0
- UniGene ID
- Rn.54471
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022274.1 |
NP_071610.1
|
2, 3 |
217 |
|
GenBank mRNA |
AF276775.1 |
- |
2, 3 |
217 |
|
View Full Product Details