Gene Transcripts

Gene Symbol
Slc22a4
Entrez Gene ID
64037
Gene Name
solute carrier family 22 (organic cation/zwitterion transporter), member 4
Gene Aliases
Octn1
Chromosome Location
Chr. 10: 39334972 - 39373508 on Build Rnor_6.0
UniGene ID
Rn.163093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022270.1 NP_071606.1 942
GenBank mRNA AF169831.1 - 942

View Full Product Details