Gene Transcripts

Gene Symbol
Rims1
Entrez Gene ID
84556
Gene Name
regulating synaptic membrane exocytosis 1
Gene Aliases
RIM1a, Rim1
Chromosome Location
Chr. 9: 28440408 - 28973246 on Build Rnor_6.0
UniGene ID
Rn.10799
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052829.1 NP_439894.1 1, 2 575
XM_008766936.1 XP_008765158.1 1, 2 575
GenBank mRNA AF007836.1 - 1, 2 575
AF199333.1 - 1, 2 575

View Full Product Details