Gene Transcripts

Gene Symbol
MTF2
Entrez Gene ID
22823
Gene Name
metal response element binding transcription factor 2
Gene Aliases
M96, PCL2, TDRD19A, dJ976O13.2
Chromosome Location
Chr. 1: 93079219 - 93139081 on Build GRCh38
UniGene ID
Hs.31016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164391.1 NP_001157863.1 2, 3 322
NM_001164392.1 NP_001157864.1 2 402
NM_007358.3 NP_031384.1 2 402
XM_011541016.1 XP_011539318.1 2 354
XM_011541017.2 XP_011539319.1 2 418
XM_011541019.1 XP_011539321.1 2, 3 274
XM_017000677.1 XP_016856166.1 2 354
XM_017000678.1 XP_016856167.1 2, 3 311
XM_017000679.1 XP_016856168.1 2 354
GenBank mRNA AF072814.1 - 1 96
AF086908.1 - 2 327
AJ010014.1 - 2 353
AK290318.1 - 2 354
AK302776.1 - 2, 3 261
BC001897.1 - 2 321
BC010013.2 - 2 326 4308565
DB173315.1 - 2 402

View Full Product Details