Gene Transcripts

Gene Symbol
Maged2
Entrez Gene ID
113947
Gene Name
melanoma antigen, family D, 2
Gene Aliases
Chromosome Location
Chr. 5: 33174539 - 33182715 on Build Rnor_6.0
UniGene ID
Rn.198407
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271109.1 NP_001258038.1 13 2013
NM_001271110.1 NP_001258039.1 13 2041
NM_080479.2 NP_536727.2 12 2162
XM_006237924.2 XP_006237986.1 13 2103
GenBank mRNA AJ293617.1 - 12 1954
AY724507.1 - 13 2026
BC086998.1 - 11 2970
BC107467.1 - 12 2162 7459969
FQ229095.1 - 13 2025

View Full Product Details