Gene Transcripts

Gene Symbol
Gmfg
Entrez Gene ID
113940
Gene Name
glia maturation factor, gamma
Gene Aliases
Chromosome Location
Chr. 1: 85312473 - 85322999 on Build Rnor_6.0
UniGene ID
Rn.15333
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181091.2 NP_851605.1 4 228
GenBank mRNA AB007364.1 - 4 228

View Full Product Details