Gene Transcripts

Gene Symbol
Capn2
Entrez Gene ID
29154
Gene Name
calpain 2, (m/II) large subunit
Gene Aliases
Chromosome Location
Chr. 13: 100878649 - 100928811 on Build Rnor_6.0
UniGene ID
Rn.6822
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017116.2 NP_058812.1 4 926
GenBank mRNA BC065306.1 - 4 926
L09120.1 - 4 612

View Full Product Details