Gene Transcripts
- Gene Symbol
- Cyp11a1
- Entrez Gene ID
- 29680
- Gene Name
- cytochrome P450, family 11, subfamily a, polypeptide 1
- Gene Aliases
- Cyp11a, Cypxia1, P450scc
- Chromosome Location
- Chr. 8: 62798342 - 62809674 on Build Rnor_6.0
- UniGene ID
- Rn.1401
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC089100.1 |
- |
9 |
1763 |
|
J05156.1 |
- |
9 |
1758 |
|
M22615.1 |
- |
4 |
709 |
|
View Full Product Details