Gene Transcripts

Gene Symbol
Galnt10
Entrez Gene ID
170501
Gene Name
polypeptide N-acetylgalactosaminyltransferase 10
Gene Aliases
Galnt9
Chromosome Location
Chr. 10: 43067316 - 43208992 on Build Rnor_6.0
UniGene ID
Rn.60224
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130742.1 NP_570098.1 2 207
GenBank mRNA AF241241.1 - 2 207

View Full Product Details