Gene Transcripts

Gene Symbol
LAMTOR2
Entrez Gene ID
28956
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
Gene Aliases
ENDAP, HSPC003, MAPBPIP, MAPKSP1AP, ROBLD3, Ragulator2, p14
Chromosome Location
Chr. 1: 156054726 - 156058510 on Build GRCh38
UniGene ID
Hs.632483
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145264.1 NP_001138736.1 3 551
NM_014017.3 NP_054736.1 4 641
GenBank mRNA AF070659.1 - 4 553
AK307086.1 - 3 477
BC024190.2 - 4 517 2819969
BU184679.1 - 3 463
DB566869.1 - 3 423

View Full Product Details