Gene Transcripts

Gene Symbol
Ciao1
Entrez Gene ID
29231
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
Wdr39
Chromosome Location
Chr. 3: 119671778 - 119677324 on Build Rnor_6.0
UniGene ID
Rn.108832
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008766.1 NP_001008766.1 5 657
GenBank mRNA BC088474.1 - 5 657
FQ230934.1 - 5 671
FQ231672.1 - 5 650

View Full Product Details