Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318234.1 | NP_001305163.1 | 7 | 1287 | |
NM_014723.3 | NP_055538.2 | 6 | 710 | ||
XM_005260889.3 | XP_005260946.1 | 6 | 808 | ||
XM_011529402.2 | XP_011527704.1 | 7 | 1385 | ||
XM_011529403.2 | XP_011527705.1 | 6 | 1357 | ||
XM_011529404.2 | XP_011527706.1 | 6 | 860 | ||
GenBank mRNA | AB002372.1 | - | 7 | 1223 | |
AF187733.1 | - | 7 | 1223 | ||
BC035788.1 | - | 6 | 685 | 5727912 |