Gene Transcripts

Gene Symbol
Vom1r46
Entrez Gene ID
494286
Gene Name
vomeronasal 1 receptor 46
Gene Aliases
V1rg15
Chromosome Location
Chr. 1: 67064847 - 67065797 on Build Rnor_6.0
UniGene ID
Rn.129421
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008939.1 NP_001008939.1 1 401
GenBank mRNA AY510348.1 - 1 401

View Full Product Details