Gene Transcripts

Gene Symbol
Pyroxd2
Entrez Gene ID
309381
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Gene Aliases
RGD1303232
Chromosome Location
Chr. 1: 261961164 - 261987033 on Build Rnor_6.0
UniGene ID
Rn.102445
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004261.1 NP_001004261.1 16 1900
XM_006231394.2 XP_006231456.1 17 2222
XM_008760424.1 XP_008758646.1 16 2489
XM_008760425.1 XP_008758647.1 16 2252
GenBank mRNA BC079368.1 - 16 1900

View Full Product Details