Gene Transcripts

Gene Symbol
TXNDC8
Entrez Gene ID
255220
Gene Name
thioredoxin domain containing 8
Gene Aliases
SPTRX-3, TRX6, bA427L11.2
Chromosome Location
Chr. 9: 110300900 - 110337887 on Build GRCh38
UniGene ID
Hs.147064
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003936.3 NP_001003936.1 6 498
NM_001286946.1 NP_001273875.1 5 438
GenBank mRNA AF305830.1 - 6 438

View Full Product Details