Gene Transcripts

Gene Symbol
TXNDC8
Entrez Gene ID
255220
Gene Name
thioredoxin domain containing 8
Gene Aliases
SPTRX-3, TRX6, bA427L11.2
Chromosome Location
Chr. 9: 110300900 - 110337887 on Build GRCh38
UniGene ID
Hs.147064
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003936.3 NP_001003936.1 4, 5 344
NM_001286946.1 NP_001273875.1 3, 4 284
XM_017014579.1 XP_016870068.1 4, 5 347
XM_017014581.1 XP_016870070.1 4, 5 347
XM_017014582.1 XP_016870071.1 3, 4 287
XM_017014584.1 XP_016870073.1 4, 5 463
GenBank mRNA AF305830.1 - 4, 5 284
BC130549.1 - 4, 5 274 40146580
BC130551.1 - 4, 5 274 40146582
BC144461.1 - 3, 4 214 9052987

View Full Product Details