Gene Transcripts

Gene Symbol
SLC39A14
Entrez Gene ID
23516
Gene Name
solute carrier family 39 member 14
Gene Aliases
HMNDYT2, LZT-Hs4, NET34, ZIP14, cig19
Chromosome Location
Chr. 8: 22367249 - 22434129 on Build GRCh38
UniGene ID
Hs.491232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128431.2 NP_001121903.1 9 3778
NM_001135153.1 NP_001128625.1 9 3819
NM_015359.4 NP_056174.2 9 3778
XM_005273465.2 XP_005273522.1 10 3866
XM_005273466.4 XP_005273523.1 9 3850
XM_006716323.2 XP_006716386.1 11 4064
XM_006716324.2 XP_006716387.1 10 3855
XM_011544478.2 XP_011542780.1 10 3930
XM_017013292.1 XP_016868781.1 10 3946
XM_017013293.1 XP_016868782.1 11 4064
XM_017013294.1 XP_016868783.1 11 3973
XM_017013295.1 XP_016868784.1 9 3983
GenBank mRNA AK172810.1 - 9 3748
BX648538.1 - 2 2132
D31887.1 - 9 3722

View Full Product Details